Would you like to log in again?
Class Apparatus Sets
Supplier: MASTERDATA QL HOLD 2
6903148
This apparatus set conforms to the 1994 Sixth Edition. The Class apparatus set recommended for 30 students has 6 additional spectroscopes, 6 additional black boxes including rods, washers, and rubber bands, and 2 additional dice sets. 1 Cloud Chamber Set (64712) 1 Molecular Size and Mass Set (64735) 1 Spectral Analysis Set (64799 or 64799-30 ) 1 Black Box Set (64796-30 ) 1 Dice Sets (Pkg. of 12) (64724) 1 Portable Geiger Counter (64331-00 ) 1 Charger, 1A6/12 Volts (45710-00 )
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Class Apparatus Set, 30 Student, IPS 6th Edition 6903148 470189-430 Kit of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty