Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Classic North American Rock Collection

Supplier: VWR
WARD470025-312
A comprehensive reference set of Petrology classes.

One hundred examples of the most common types of igneous, sedimentary, and metamorphic rocks from North American localities show the distribution of different rock types throughout the continent. The large sample size provides easy viewing of individual characteristics. The included 108-page manual contains detailed information, including microscopic and megascopic descriptions, for each specimen, plus name, locality, and geologic background. The set is housed in individual, labeled plastic trays.

Contents:100 numbered samples, 3" x 4". Rocks: Igneous (43), Sedimentary (27), Metamorphic (30).

Classic North American Rock Collection, Student Set contains the same 100 rocks as in the Classic North American Rock Collection, in a smaller sample size ideal for individual study or for making thin sections. The set is housed in two fifty-compartment collection boxes and an identification list is included.

Contents: 100 numbered samples, 1" x 2".

Contents: 100 numbered thin section glass microscope slides, 27 x 46 mm.
Classic North American Rock Collection with Matching Thin Sections is a complete set of specimens and slides at a special value. The same 100 rocks as in the Classic North American Rock Collection, plus 100 thin sections corresponding to each rock.
Contents: 100 numbered samples, 3" x 4", 100 numbered thin section glass microscope slides, 27 x 46 mm.



 

ORDER

Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
Classic North American Rock Collection   470025-312 Each Retrieving Restricted
restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty