Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Denoyer-Geppert® Giant DNA Molecule Model

Supplier: DENOYER-GEPPERT SCI CO SE
0611-00
This extra-large model can be seen anywhere in the classroom.

  • Model Integrates Biology, Physics, and Math
  • Extra-Large Model
  • Model Mounted on Rotatable Wooden Stand
  • Color-Coded Atoms
  • Instruction Manual Included

Teachers can use the model for demonstrations or have students participate in hands-on study of the DNA double helix. Either way, students can view the details of the large model regardless of where they’re sitting.

It's easy to unzip the DNA to recreate RNA synthesis as the parts are linked to detachable magnetic connectors. Atoms are color coded for easy identification. The model is mounted on a rotatable wooden stand and comes with an instruction manual.



 

ORDER

Expand All / Collapse All
Description Height Length Width Supplier No. VWR Catalog Number Unit Price Quantity
Giant DNA Molecule 86 cm 41 cm 41 cm 0611-00 470004-756 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping