ContactUsLinkComponent
Growing Gourmet Mushrooms Lab Activity
Growing Gourmet Mushrooms Lab Activity
Catalog #: 470193-166
Growing Gourmet Mushrooms Lab Activity
Catalog #: 470193-166
Price
87,20 $
Log in for availability
Unit of Measure Kit of 1
Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Specifications
  • Description:
    Growing Gourmet Mushrooms Lab Activity
  • Cat. no.:
    470193-166
Specifications
About this item
Grow Exotic-Looking Mushrooms
Present a comprehensive unit on fungi by growing oyster mushrooms and watching them transform sawdust into nutritious soil. Your students will learn the importance of fungi, how they reproduce, and the structure and function of all the parts. In addition, they’ll investigate the effects of gravity (geotropism) on mushroom growth adaptive responses and the implication for fungal reproduction. The kit contains one control and one treatment setup, and literature with correlation to the national science standards. Note: Includes perishable materials. Please specify delivery date. Only shipped on Mondays and Tuesdays.
Customers Who Bought This Also Bought