Leslie’s Cube
Supplier: VWR International
PH0736
470017-204EA 51 CAD
470017-204
Leslie’s Cube
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Illustrate That Surfaces At the Same Temperature Do Not Radiate Equally.
  • Each of the four sides have different surfaces (bare metal, white, flat black, and gloss black) and the top has an opening for a thermometer
  • The large 6" sides are ideal for classroom demonstration