Salinity
Supplier: LA MOTTE CHEMICAL PROD CO SE
R-7459-01
470021-206EA 78.6 CAD
470021-206
Salinity
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Precalibrated test tubes and labware give fast, accurate results on-site, in the classroom, or in the lab. Reagents, labware, and step-by-step instructions are furnished in a compact, waterproof carrying case. Kit contains materials for 50 tests.
Range: 0–40 ppt.