Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cogent F5™, PentaFluroPhenyl 60Å, HPLC Columns, MicroSolv

Supplier: MicroSolv Technology Corp
60322-15P-2 60022-02 60022-01-R 60022-25P 60022-02R-2
Cogent F5™ HPLC Columns are designed to provide a unique separation mechanism for a wide range of molecules but are particularly effective for retention of fluorinated compounds

Using an Acetonitrile and Water mobile phase, aromatic compounds (commonly found in pharmaceuticals) are easily separated. When used with fluorinated solvents such as 2,2,2-trifluoroethanol, a very useful selectivity for compounds will be observed for hydrogen bonding/donor functionalities. Using Trifluoracetic acid at low pH can be effective for peptide and other biological separations.

An increased retention may be found compared with Phenyl, C8, C18, or other alkyl based columns and may initiate reversal in retention order in natural products such as Taxol™ or taxanes.

Cogent F5™ is very useful to differentiate between halogen bearing compounds and their analogs and positional isomers




 

ORDER

Expand All / Collapse All
Description Column Format I.D. x L Particle Size Supplier No. VWR Catalog Number Unit Price Quantity
Column Analytical 2.1 x 150 mm 3 µm 60322-15P-2 97051-028 Each Retrieving
Column Analytical 4.0 x 20 mm 5 µm 60022-01-R 97051-022 Pack of 5 Retrieving
Column Analytical 2.0 x 20 mm 5 µm 60022-02R-2 97051-026 Pack of 5 Retrieving
Column Analytical 4.6 x 250 mm 5 µm 60022-25P 97057-738 Each Retrieving
Column with Holder Analytical 4.0 x 20 mm 5 µm 60022-02 97051-024 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty