Would you like to log in again?
Universal Digital Microscope Stand, Aven Inc.
Supplier: AVEN TOOLS INC.
26700-311
This compact universal stand allows for precise focusing and measurement.
  • Built-in X-Y Stage offers precise fine adjustment
  • Fits Mighty Scopes 1.3M and 5M, and ZipScopes
An adapter enables use with a variety of digital microscopes. The built-in LED backlight is powered via USB or 4 AAA batteries. Features metal clips for holding objects in place while being viewed. Glass viewing platform is 40×40 mm.
ORDER
Expand All / Collapse All
Description Base Dimensions Dimensions Supplier No. VWR Catalog Number Unit Price Quantity
Universal Digital Microscope Stand 120×110 mm 11L×12W×18H cm (41/3×43/4×7") 26700-311 89239-410 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty