1-Butoxy-4-nitrobenzene ≥99.0%

Supplier: TCI America

Synonyms: Butyl 4-Nitrophenyl Ether

B1162-25G
TCB1162-025GEA 327.06 CAD
TCB1162-025G
1-Butoxy-4-nitrobenzene ≥99.0%
1-Butoxy-4-nitrobenzene

CAS Number: 7244-78-2
MDL Number: MFCD00024688
Molecular Formula: C10H13NO3
Molecular Weight: 195.22
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Freezing point (°C): 31


Other Product Names / Synonyms: Butyl 4-Nitrophenyl Ether

Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
MDL Number: MFCD00024688
CAS Number: 7244-78-2
https://ca.vwr.com/stibo/web/std.lang.all/30/05/28933005.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR