1-Benzyl-3-(tert-butoxycarbonylamino)pyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: tert-Butyl (1-benzylpyrrolidin-3-yl)carbamatetert-butyl N-(1-benzylpyrrolidin-3-yl)carbamate

B1357-10G
TCB1357-010GEA 85.32 CAD
TCB1357-010G
1-Benzyl-3-(tert-butoxycarbonylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
1-Benzyl-3-(tert-butoxycarbonylamino)pyrrolidine

CAS Number: 99735-30-5
MDL Number: MFCD00059039
Molecular Formula: C16H24N2O2
Molecular Weight: 276.38
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 113

Formula: C₁₆H₂₄N₂O₂
MW: 276.38 g/mol
Melting Pt: 113 °C
MDL Number: MFCD00059039
CAS Number: 99735-30-5
https://ca.vwr.com/stibo/web/std.lang.all/31/70/28933170.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR