3-Methyl-2-butenyl benzoate ≥98.0%

Supplier: TCI America

Synonyms: Benzoic Acid 3-Methyl-2-butenyl EsterPrenyl Benzoate3-Methylbut-2-en-1-yl benzoateBenzoic Acid Prenyl Ester

B1409-25G
TCB1409-025GEA 213.3 CAD
TCB1409-025G
3-Methyl-2-butenyl benzoate ≥98.0%
3-Methyl-2-butenyl benzoate

CAS Number: 5205-11-8
MDL Number: MFCD00036570
Molecular Formula: C12H14O2
Molecular Weight: 190.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 107
Flash Point (°C): 134
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: Benzoic Acid 3-Methyl-2-butenyl Ester, Benzoic Acid Prenyl Ester, Prenyl Benzoate

Formula: C₁₂H₁₄O₂
MW: 190.24 g/mol
Boiling Pt: 107 °C
Density: 1.03 (20 °C)
Flash Pt: 134 °C
MDL Number: MFCD00036570
CAS Number: 5205-11-8
https://ca.vwr.com/stibo/web/std.lang.all/32/09/28933209.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR