(+)-Benzylmethylphenylsilylacetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B1720-100MG
TCB1720-100MGEA 276 CAD
TCB1720-100MG
(+)-Benzylmethylphenylsilylacetic acid ≥98.0% (by titrimetric analysis)
(+)-Benzylmethylphenylsilylacetic acid

[for e.e. Determination by NMR]
CAS Number: 95373-54-9
Molecular Formula: C16H18O2Si
Molecular Weight: 270.40
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 70
Specific rotation [a]20/D: 7 deg (C=2, CHCl3)

Formula: C₁₆H₁₈O₂Si
Melting Pt: 70 °C
CAS Number: 95373-54-9
https://ca.vwr.com/stibo/web/std.lang.all/34/71/28933471.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR