6-Bromobenzo[d]thiazol-2(3H)-one ≥97.0%

Supplier: TCI America

Synonyms: 6-Bromo-2-benzothiazolinone

B2012-1G
TCB2012-001GEA 104.28 CAD
TCB2012-001G
6-Bromobenzo[d]thiazol-2(3H)-one ≥97.0%
6-Bromobenzo[d]thiazol-2(3H)-one

CAS Number: 62266-82-4
MDL Number: MFCD00239363
Molecular Formula: C7H4BrNOS
Molecular Weight: 230.08
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 230

Formula: C₇H₄BrNOS
MW: 230.08 g/mol
Melting Pt: 230 °C
MDL Number: MFCD00239363
CAS Number: 62266-82-4
https://ca.vwr.com/stibo/web/std.lang.all/37/71/28933771.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR