1,1-Bis(3-cyclohexyl-4-hydroxyphenyl)cyclohexane ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4,4'-Cyclohexylidenebis(2-cyclohexylphenol)

B2752-25G
TCB2752-25GEA 327.06 CAD
TCB2752-25G
1,1-Bis(3-cyclohexyl-4-hydroxyphenyl)cyclohexane ≥98.0% (by HPLC)
1,1-Bis(3-cyclohexyl-4-hydroxyphenyl)cyclohexane

CAS Number: 4221-68-5
Molecular Formula: C30H40O2
Molecular Weight: 432.65
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 166


Product Categories: Materials Chemistry, Monomers, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: 4,4'-Cyclohexylidenebis(2-cyclohexylphenol)

Formula: C₃₀H₄₀O₂
Melting Pt: 166 °C
CAS Number: 4221-68-5
https://ca.vwr.com/stibo/web/std.lang.all/42/78/28934278.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR