1-Bromo-2-fluorocyclohexane ≥96.0%

Supplier: TCI America

B3053-1G
TCB3053-1GEA 449.82 CAD
TCB3053-1G
1-Bromo-2-fluorocyclohexane ≥96.0%
1-Bromo-2-fluorocyclohexane

CAS Number: 656-57-5
MDL Number: MFCD01320789
Molecular Formula: C6H10BrF
Molecular Weight: 181.05
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 78
Specific Gravity (20/20): 1.46

Formula: C₆H₁₀BrF
Boiling Pt: 78 °C
Density: 1.46 (20 °C)
MDL Number: MFCD01320789
CAS Number: 656-57-5
https://ca.vwr.com/stibo/web/std.lang.all/44/97/28934497.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR