2-(4-Chlorophenyl)-2-methylpropanol ≥98.0%

Supplier: TCI America

C2176-5G
TCC2176-5GEA 150.1 CAD
TCC2176-5G
2-(4-Chlorophenyl)-2-methylpropanol ≥98.0%
2-(4-Chlorophenyl)-2-methylpropanol

CAS Number: 80854-14-4
MDL Number: MFCD06657569
Molecular Formula: C10H13ClO
Molecular Weight: 184.66
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 50

Formula: C₁₀H₁₃ClO
Melting Pt: 50 °C
MDL Number: MFCD06657569
CAS Number: 80854-14-4
https://ca.vwr.com/stibo/web/std.lang.all/91/97/28939197.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR