2,3-Dichloro-5-nitro-1,4-naphthoquinone ≥97.0%

Supplier: TCI America

D2428-5G
TCD2428-005GEA 357.08 CAD
TCD2428-005G
2,3-Dichloro-5-nitro-1,4-naphthoquinone ≥97.0%
2,3-Dichloro-5-nitro-1,4-naphthoquinone

CAS Number: 22360-86-7
MDL Number: MFCD00192929
Molecular Formula: C10H3Cl2NO4
Molecular Weight: 272.04
Purity/Analysis Method: >97.0% (GC)
Form: Crystal

Formula: C₁₀H₃Cl₂NO₄
MW: 272.04 g/mol
MDL Number: MFCD00192929
CAS Number: 22360-86-7
https://ca.vwr.com/stibo/web/std.lang.all/93/55/28979355.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR