1,2,10-Decanetriol ≥97.0%

Supplier: TCI America

Synonyms: 1,2,10-Trihydroxydecane

D2721-5G
TCD2721-005GEA 466.1 CAD
TCD2721-005G
1,2,10-Decanetriol ≥97.0%
1,2,10-Decanetriol

CAS Number: 91717-85-0
MDL Number: MFCD00797600
Molecular Formula: C10H22O3
Molecular Weight: 190.28
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 66


Other Product Names / Synonyms: 1,2,10-Trihydroxydecane

Formula: C₁₀H₂₂O₃
MW: 190.28 g/mol
Melting Pt: 66 °C
MDL Number: MFCD00797600
CAS Number: 91717-85-0
https://ca.vwr.com/stibo/web/std.lang.all/98/39/28979839.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR