Diethyl-2-chloro-3-oxosuccinate ≥93.0%

Supplier: TCI America

Synonyms: Diethyl 2-chloro-3-oxosuccinateChlorooxalacetic acid diethyl esterDiethyl chlorooxalacetateDiethyl 2-chlorooxalacetate

D3635-1G
TCD3635-1GEA 162.74 CAD
TCD3635-1G
Diethyl-2-chloro-3-oxosuccinate ≥93.0%
Diethyl-2-chloro-3-oxosuccinate

CAS Number: 34034-87-2
MDL Number: MFCD04114359
Molecular Formula: C8H11ClO5
Molecular Weight: 222.62
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Specific Gravity (20/20): 1.23
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Chlorooxalacetic Acid Diethyl Ester

Formula: C₈H₁₁ClO₅
MW: 222.6229 g/mol
Boiling Pt: 130 °C
Density: 1.23 (20 °C)
MDL Number: MFCD04114359
CAS Number: 34034-87-2
https://ca.vwr.com/stibo/web/std.lang.all/56/24/28985624.jpg

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR