1-Hydroxyphenazine ≥95.0%

Supplier: TCI America

H0289-100MG H0289-5G H0289-1G
TCH0289-100MGEA 133.5 CAD
TCH0289-100MG TCH0289-5G TCH0289-1G
1-Hydroxyphenazine ≥95.0%
1-Hydroxyphenazine

CAS Number: 528-71-2
MDL Number: MFCD00059692
Molecular Formula: C12H8N2O
Molecular Weight: 196.21
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 158

Formula: C₁₂H₈N₂O
Melting Pt: 158 °C
MDL Number: MFCD00059692
CAS Number: 528-71-2
https://ca.vwr.com/stibo/web/std.lang.all/03/33/28970333.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR