(1H-Benzoimidazol-2-yl)methanol ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (2-Benzimidazolyl)methanol2-(Hydroxymethyl)benzimidazole2-Benzimidazolemethanol

H1229-5G
TCH1229-5GEA 235.42 CAD
TCH1229-5G
(1H-Benzoimidazol-2-yl)methanol ≥98.0% (by HPLC, titration analysis)
(1H-Benzoimidazol-2-yl)methanol

CAS Number: 4856-97-7
MDL Number: MFCD00014560
Molecular Formula: C8H8N2O
Molecular Weight: 148.17
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 172


Other Product Names / Synonyms: 2-Benzimidazolemethanol, (2-Benzimidazolyl)methanol

Formula: C₈H₈N₂O
MW: 148.1619 g/mol
Melting Pt: 172 °C
MDL Number: MFCD00014560
CAS Number: 4856-97-7
https://ca.vwr.com/stibo/web/std.lang.all/11/56/28971156.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR