2-(Isobutylthio)ethanol ≥97.0%

Supplier: TCI America

Synonyms: 2-Hydroxyethyl Isobutyl Sulphide2-Hydroxyethyl Isobutyl Sulfide

I0264-5ML
TCI0264-005MLEA 249.39 CAD
TCI0264-005ML
2-(Isobutylthio)ethanol ≥97.0%
2-(Isobutylthio)ethanol

CAS Number: 42779-10-2
MDL Number: MFCD00014040
Molecular Formula: C6H14OS
Molecular Weight: 134.24
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: 2-Hydroxyethyl Isobutyl Sulfide

Formula: C₆H₁₄OS
Boiling Pt: 88 °C
Density: 0,97 (20 °C)
MDL Number: MFCD00014040
CAS Number: 42779-10-2

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR