7-Octenyl acetate ≥98.0%
Supplier: TCI America
A1501-25G
A1501-5G
TCA1501-025GEA
774.2
CAD
TCA1501-025G
TCA1501-005G
7-Octenyl acetate ≥98.0%
7-Octenyl acetate
CAS Number: 5048-35-1
MDL Number: MFCD01861278
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Specific Gravity (20/20): 0.89
Other Product Names / Synonyms: 8-Acetoxy-1-octene, Acetic Acid 7-Octenyl Ester
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...