2-[(4-Methoxybenzylidene)amino]fluorene ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-[(p-Anisylidene)amino]fluorene2-(p-Anisalamino)fluorene

M0586-5G
TCM0586-005GEA 532.46 CAD
TCM0586-005G
2-[(4-Methoxybenzylidene)amino]fluorene ≥98.0% (by titrimetric analysis)
2-[(4-Methoxybenzylidene)amino]fluorene

CAS Number: 5424-78-2
MDL Number: MFCD00019064
Molecular Formula: C21H17NO
Molecular Weight: 299.37
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 2-(p-Anisalamino)fluorene, 2-[(p-Anisylidene)amino]fluorene

Formula: C₂₁H₁₇NO
MW: 299.37 g/mol
MDL Number: MFCD00019064
CAS Number: 5424-78-2
https://ca.vwr.com/stibo/web/std.lang.all/40/44/28974044.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR