(2-Methylbutyryl)shikonin ≥88.0% (by HPLC)

Supplier: TCI America

Synonyms: 5,8-Dihydroxy-2-[1-(2-methylbutyryloxy)-4-methyl-3-pentenyl]-1,4-naphthalenedione

M1028-100MG
TCM1028-100MGEA 158 CAD
TCM1028-100MG
(2-Methylbutyryl)shikonin ≥88.0% (by HPLC)
(2-Methylbutyryl)shikonin

CAS Number: 52387-15-2
MDL Number: MFCD00142946
Molecular Formula: C21H24O6
Molecular Weight: 372.42
Purity/Analysis Method: >88.0% (HPLC)
Form: Paste


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Others (Phytochemicals)
Other Product Names / Synonyms: 5,8-Dihydroxy-2-[1-(2-methylbutyryloxy)-4-methyl-3-pentenyl]-1,4-naphthalenedione

Formula: C₂₁H₂₄O₆
MDL Number: MFCD00142946
CAS Number: 52387-15-2
https://ca.vwr.com/stibo/web/std.lang.all/46/45/28974645.jpg

Order Now

Specification Test Results

Purity/Analysis Method 88.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR