(R)-(+)-4-(Methoxymethyl)-1,3-dioxolan-2-one ≥98.0%

Supplier: TCI America

Synonyms: (R)-(+)-4-(Methoxymethyl)-2-oxo-1,3-dioxolane(R)-(+)-3-Methoxypropylene Carbonate

M1451-25G M1451-5G
TCM1451-5GEA 205.4 CAD
TCM1451-5G TCM1451-25G
(R)-(+)-4-(Methoxymethyl)-1,3-dioxolan-2-one ≥98.0%
(R)-(+)-4-(Methoxymethyl)-1,3-dioxolan-2-one

CAS Number: 185836-34-4
MDL Number: MFCD01321303
Molecular Formula: C5H8O4
Molecular Weight: 132.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Flash Point (°C): 110
Specific Gravity (20/20): 1.24
Specific rotation [a]20/D: 46 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(+)-4-(Methoxymethyl)-2-oxo-1,3-dioxolane, (R)-(+)-3-Methoxypropylene Carbonate

Formula: C₅H₈O₄
Boiling Pt: 106 °C
Density: 1.24 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD01321303
CAS Number: 185836-34-4
https://ca.vwr.com/stibo/web/std.lang.all/56/35/28975635.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR