4-Nitroaniline hydrochloride ≥99.0% (by HPLC, titration analysis)
Supplier: TCI America
N0122-25G
TCN0122-025GEA
466.1
CAD
TCN0122-025G
4-Nitroaniline hydrochloride ≥99.0% (by HPLC, titration analysis)
4-Nitroaniline hydrochloride
CAS Number: 15873-51-5
MDL Number: MFCD00054338
Molecular Formula: C6H6N2O2
Molecular Weight: 174.58
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Formula:
C₆H₇ClN₂O₂ MW: 174.5850 g/mol |
MDL Number:
MFCD00054338 CAS Number: 15873-51-5 UN: 1661 ADR: 6.1,II |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...