4-Methylhippuric acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

T0313-25G T0313-5G
TCT0313-025GEA 293.76 CAD
TCT0313-025G TCT0313-5G
4-Methylhippuric acid ≥98.0% (by HPLC, titration analysis)
4-Methylhippuric acid

CAS Number: 27115-50-0
MDL Number: MFCD00020449
Molecular Formula: C10H11NO3
Molecular Weight: 193.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 161


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-(4-Methylbenzoyl)glycine, 4-Methylhippuric Acid

https://ca.vwr.com/stibo/web/std.lang.all/46/05/28994605.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR