3-(Trimethylsilylethynyl)pyridine ≥98.0%

Supplier: TCI America

Synonyms: 3-((Trimethylsilyl)ethynyl)pyridine

T1963-1G
TCT1963-1GEA 295.46 CAD
TCT1963-1G
3-(Trimethylsilylethynyl)pyridine ≥98.0%
3-(Trimethylsilylethynyl)pyridine

CAS Number: 80673-00-3
MDL Number: MFCD01863744
Molecular Formula: C10H13NSi
Molecular Weight: 175.31
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 94
Specific Gravity (20/20): 0.94
Storage Temperature: <0°C


Other Product Names / Synonyms: (3-Pyridylethynyl)trimethylsilane, 1-(3-Pyridyl)-2-trimethylsilylacetylene

Formula: C₁₀H₁₃NSi
MW: 175.31 g/mol
Density: 0.94 (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD01863744
CAS Number: 80673-00-3
https://ca.vwr.com/stibo/web/std.lang.all/59/15/28995915.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR