2,3,4,6-Tetra-O-benzyl-D-glucopyranosyl fluoride ≥96.0% (by HPLC)

Supplier: TCI America

T1971-500MG
TCT1971-500MGEA 594.08 CAD
TCT1971-500MG
2,3,4,6-Tetra-O-benzyl-D-glucopyranosyl fluoride ≥96.0% (by HPLC)
2,3,4,6-Tetra-O-benzyl-D-glucopyranosyl fluoride

CAS Number: 122741-44-0
MDL Number: MFCD01862264
Molecular Formula: C34H35FO5
Molecular Weight: 542.65
Purity/Analysis Method: >96.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 45
Specific rotation [a]20/D: 28 deg (C=1, CHCl3)
Storage Temperature: 0-10°C

Formula: C₃₄H₃₅FO₅
MW: 542.65 g/mol
Melting Pt: 45 °C
MDL Number: MFCD01862264
CAS Number: 122741-44-0
https://ca.vwr.com/stibo/web/std.lang.all/59/22/28995922.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR