2,3,4,6-Tetra-O-pivaloyl-D-mannopyranosyl fluoride ≥95.0%

Supplier: TCI America

T2568-1G
TCT2568-1GEA 491.38 CAD
TCT2568-1G
2,3,4,6-Tetra-O-pivaloyl-D-mannopyranosyl fluoride ≥95.0%
2,3,4,6-Tetra-O-pivaloyl-D-mannopyranosyl fluoride

Molecular Weight: 518.62
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Storage Temperature: 0-10°C
Molecular Formula: C26H43FO9

Formula: C₂₆H₄₃FO₉
MW: 518.62 g/mol
MDL Number: MFCD11035863
CAS Number: 187269-63-2
https://ca.vwr.com/stibo/web/std.lang.all/62/84/28996284.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR