Rhodium(III) oxide, anhydrous ≥99.9% (metals basis) Rh ≥ 80.6%

Supplier: THERMO FISHER SCIENTIFIC CA

11814-04 11814-02 11814-04 11814-02
CAAA11814-04EA 2240.85 CAD
AA11814-04 AA11814-02 CAAA11814-04 CAAA11814-02
Rhodium(III) oxide, anhydrous ≥99.9% (metals basis) Rh ≥ 80.6%
Rhodium (III) oxide
Formula: O₃Rh₂
Melting Pt: 1100 °C (dec.)
Density: 8.20
Storage Temperature: Ambient
MDL Number: MFCD00011205
CAS Number: 12036-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR