Graphite ink for tantalum capacitors
Supplier: Thermo Scientific Chemicals
Warning
42193-A1 42193-30 42193-30 42193-A1
CAAA42193-30EA 224.53 CAD
AA42193-A1 CAAA42193-30 CAAA42193-A1 AA42193-30
Graphite ink for tantalum capacitors
Graphite ink
A graphite ink for use as an undercoat for silver ink in the production of tantalum capacitors