Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 0.20 micron
Supplier: Thermo Scientific Chemicals
42713-4Y
42713-4Y
CAAA42713-4YEA
736.63
CAD
AA42713-4Y
CAAA42713-4Y
Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 0.20 micron
Poly(styrene)
Monodisperse latex spheres stabilized with a slight anionic charge from surface sulfate groups
Formula:
(C₈H₈)n MW: 125,000-250,000 Storage Temperature: Refrigerator |
MDL Number:
MFCD00084450 CAS Number: 9003-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...