3-Amino-4-pyrazolecarbonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-1H-pyrazole-4-carbonitrile5-Aminopyrazole-4-carbonitrile3-Amino-4-cyanopyrazole

A14266-03 A14266-06 A14266-14 A14266-06 A14266-03
CAAAA14266-06EA 142.8 CAD
AAAA14266-14 CAAAA14266-06 AAA14266-03 CAAAA14266-03 AAA14266-06
3-Amino-4-pyrazolecarbonitrile 98%
3-Amino-4-pyrazolecarbonitrile
Formula: C₄H₄N₄
MW: 108.10 g/mol
Melting Pt: 173…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00005237
CAS Number: 16617-46-2
EINECS: 240-665-6
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR