Phenylphosphinic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

A17453-36 A17453-36
CAAAAA17453-36EA 192.22 CAD
CAAAAA17453-36 AAAA17453-36
Phenylphosphinic acid 98%
Phenylphosphinic acid
Formula: C₆H₇O₂P
Melting Pt: 83…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00002131
CAS Number: 1779-48-2
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR