4-Methoxy-3-nitrobenzamide 97%

Supplier: Thermo Scientific

Warning

B24905-14 B24905-03 B24905-14 B24905-03 B24905-06 B24905-06
CAAAAB24905-14EA 1183.8 CAD
CAAAAB24905-14 CAAAAB24905-03 AAAB24905-14 AAAB24905-03 CAAAAB24905-06 AAAB24905-06
4-Methoxy-3-nitrobenzamide 97%
4-Methoxy-3-nitrobenzamide
Formula: C₈H₈N₂O₃
Melting Pt: 173…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00017136
CAS Number: 10397-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR