2-(Benzylthio)-5-nitrobenzonitrile 97%

Supplier: Thermo Scientific

Warning

B24924-03 B24924-03 B24924-06 B24924-06
CAAAAB24924-03EA 178.41 CAD
AAAB24924-03 CAAAAB24924-03 CAAAAB24924-06 AAAB24924-06
2-(Benzylthio)-5-nitrobenzonitrile 97%
2-(Benzylthio)-5-nitrobenzonitrile
Formula: C₁₄H₁₀N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD00067808
CAS Number: 175135-67-8
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR