6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid 99%

Warning

H50313-MD H50313-MD H50313-03 H50313-03
CAAAAH50313-MDEA 201.81 CAD
AAAH50313-03 CAAAAH50313-MD CAAAAH50313-03 AAAH50313-MD
6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid 99%
6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Formula: C₈H₅ClN₂O₂
Melting Pt: 246…247 °C
Storage Temperature: Ambient
MDL Number: MFCD02186559
CAS Number: 182181-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR