1-(Pyrimidin-2-yl)piperidine-4-carboxylic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

H50351-03 H50351-MD H50351-MD H50351-03
CAAAAH50351-MDEA 109.73 CAD
AAAH50351-03 CAAAAH50351-MD AAAH50351-MD CAAAAH50351-03
1-(Pyrimidin-2-yl)piperidine-4-carboxylic acid 95%
1-(Pyrimidin-2-yl)piperidine-4-carboxylic acid
Formula: C₁₀H₁₃N₃O₂
MW: 207.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00664474
CAS Number: 303144-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR