4-Phenoxypiperidine 99%

Supplier: Thermo Scientific Chemicals
Warning

H50984-MD H50984-03 H50984-06 H50984-03 H50984-MD
AAH50984-06EA 883.85 CAD
AAH50984-06 AAAH50984-03 CAAAAH50984-MD CAAAAH50984-03 AAAH50984-MD
4-Phenoxypiperidine 99%
4-Phenoxypiperidine
Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
Boiling Pt: 107…108 °C/0.1 mm
Storage Temperature: Ambient
MDL Number: MFCD04114971
CAS Number: 3202-33-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR