2-Acetamido-5-nitrothiazole 98%

Supplier: VWR
Warning

Synonyms: N-(5-Nitro-2-thiazolyl)acetamide

L01356-06 L01356-06 L01356-03 L01356-03
CAAAAL01356-06EA 578.6 CAD
AAAL01356-03 CAAAAL01356-06 AAAL01356-06 CAAAAL01356-03
2-Acetamido-5-nitrothiazole 98%
2-Acetamidot-5-nitrohiazole
Formula: C₅H₅N₃O₃S
Melting Pt: ca 263 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00022438
CAS Number: 140-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR