N-(4-Methoxybenzylidene)aniline 98%

Warning

L01951-06 L01951-06 L01951-14 L01951-14
CAAAAL01951-14EA 278.16 CAD
AAAL01951-14 CAAAAL01951-14 AAAL01951-06 CAAAAL01951-06
N-(4-Methoxybenzylidene)aniline 98%
N-(4-Methoxybenzylidene)aniline
Formula: C₁₄H₁₃NO
MW: 211.26 g/mol
Melting Pt: 61…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00025836
CAS Number: 836-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR