Acetophenone azine 97%
Supplier: Thermo Scientific Chemicals
L03924-06
L03924-06
CAAAAL03924-06EA
207.65
CAD
AAAL03924-06
CAAAAL03924-06
Acetophenone azine 97%
Acetophenone azine
Formula:
C₁₆H₁₆N₂ Melting Pt: 120…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015011 CAS Number: 729-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...