Harmine ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Banisterine

L19068-MD L19068-03 L19068-MD L19068-03
CAAAAL19068-03EA 333.08 CAD
CAAAAL19068-03 CAAAAL19068-MD AAAL19068-03 AAAL19068-MD
Harmine ≥98%
Harmine

A monoamine oxidase inhibitor

Formula: C₁₃H₁₂N₂O
Melting Pt: 262…266 °C
Storage Temperature: Ambient
MDL Number: MFCD00004958
CAS Number: 442-51-3
EINECS: 207-131-4
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR