4-(Trifluoromethylthio)phenyl isocyanate 97%

Warning

L19206-03 L19206-03
CAAAAL19206-03EA 957.42 CAD
AAAL19206-03 CAAAAL19206-03
4-(Trifluoromethylthio)phenyl isocyanate 97%
4-(Trifluoromethylthio)phenyl isocyanate
Formula: C₈H₄F₃NOS
MW: 219.19 g/mol
Boiling Pt: 175 °C
Density: 1.385
Flash Pt: 80 °C(176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036025
CAS Number: 24032-84-6
UN: 2206
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR