4-Chloro-3-picoline hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloro-3-methylpyridine hydrochloride

L19558-03 L19558-MD L19558-03
CAAAAL19558-MDEA 67.51 CAD
CAAAAL19558-MD AAAL19558-03 CAAAAL19558-03
4-Chloro-3-picoline hydrochloride 97%
4-Chloro-3-picoline hydrochloride
Formula: C₆H₇Cl₂N
Melting Pt: ca 185 °C
Storage Temperature: Ambient
MDL Number: MFCD03092890
CAS Number: 19524-08-4
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR