2-(4-Phenoxyphenyl)ethylboronic acid diethanolamine ester 97%

Warning

L19703-MC L19703-MC
CAAAAL19703-MCEA 127.65 CAD
AAAL19703-MC CAAAAL19703-MC
2-(4-Phenoxyphenyl)ethylboronic acid diethanolamine ester 97%
2-(4-Phenoxyphenyl)ethylboronic acid diethanolamine ester
Formula: C₁₈H₂₂BNO₃
Melting Pt: 178…191 °C
Storage Temperature: Ambient
MDL Number: MFCD12546199
CAS Number: 608534-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR