3-Fluoropiperidine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

L19776-03 L19776-MD L19776-MD L19776-03
CAAAAL19776-03EA 783.21 CAD
CAAAAL19776-03 CAAAAL19776-MD AAAL19776-03 AAAL19776-MD
3-Fluoropiperidine hydrochloride 97%
3-Fluoropiperidine hydrochloride
Formula: C₅H₁₀FN·HCl
MW: 139.60 g/mol
Melting Pt: ca 227 °C
Storage Temperature: Ambient
MDL Number: MFCD03452799
CAS Number: 737000-77-0
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR