3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-((tert-Butoxycarbonyl)amino)-3-(3-methoxyphenyl)propanoic acid

L19807-MD L19807-03 L19807-MD L19807-03
CAAAAL19807-MDEA 85.37 CAD
AAAL19807-MD AAAL19807-03 CAAAAL19807-MD CAAAAL19807-03
3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid 99%
3-(Boc-amino)-3-(3-methoxyphenyl)propionic acid
Formula: C₁₅H₂₁NO₄
MW: 295.34 g/mol
Melting Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD02090693
CAS Number: 284493-53-4
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR